CAS RN 331949-35-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ Nexinhib20
Tocris Bioscience™ Nexinhib20
Rab27 inhibitor (IC50 = 330 nM); inhibits neutrophil exocytosis; active in vivo