Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nexinhib20
GREENER_CHOICESDP

Catalog No. 60895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-895 5 mg
60-892-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-895 Supplier Tocris Bioscience™ Supplier No. 6089/5
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rab27 inhibitor (IC50 = 330 nM); inhibits neutrophil exocytosis; active in vivo

Nexinhib20 is a Rab27 inhibitor (IC50 = 330 nM); inhibits Rab27a-JFC1 binding. Selectively inhibits neutrophil exocytosis, with no significant effect on other important innate immune responses including phagocytosis. Suppresses extracellular production of superoxide anion and cytochrome b plasma membrane expression. Also inhibits upregulation of adhesion molecules CD11b and CD66b. Displays in vivo anti-inflammatory activity.

Specifications

Chemical Name or Material 4,4-Dimethyl-1-(3-nitrophenyl)-2-(1H-1,2,4-triazol-1-yl)-1-penten-3-one
CAS 331949-35-0
Quantity 5 mg
Molecular Formula C15H16N4O3
Formula Weight 300.31
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.