accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33234-36-5

IUPAC Name: 2-chloro-3-methoxybenzoic acid
Synonyms: 2-chloro-3-methoxybenzoic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: VIUDWLLKFANPLX-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C(O)=O)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results