accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOOCH3

CAS RN 3386-32-1

IUPAC Name: octadecyl 4-methylbenzene-1-sulfonate
Synonyms: octadecyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 424.68
Molecular Formula: C25H44O3S
InChi Key: IZSBVVGANRHKEE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results