accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 341-69-5

IUPAC Name: hydrogen dimethyl({2-[(2-methylphenyl)(phenyl)methoxy]ethyl})amine chloride
Synonyms: hydrogen orphenadrine chloride
Molecular Weight (g/mol): 305.85
Molecular Formula: C18H24ClNO
InChi Key: UQZKYYIKWZOKKD-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)