accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34446-11-2

IUPAC Name: 1,2-dibromo-3-[(2,3-dibromopropoxy)methoxy]propane
Synonyms: 1,2-dibromo-3-[(2,3-dibromopropoxy)methoxy]propane
Molecular Weight (g/mol): 447.79
Molecular Formula: C7H12Br4O2
InChi Key: DUQWOWBBMLWHPB-UHFFFAOYNA-N
SMILES: BrCC(Br)COCOCC(Br)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results