CAS RN 345630-40-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ PTP CD45 Inhibitor
The PTP CD45 Inhibitor, also referenced under CAS 345630-40-2, controls the biological activity of PTP CD45. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
Tocris Bioscience™ SF 1670
PTEN inhibitor