CAS RN 3458-28-4
Thermo Scientific Chemicals D(+)-Mannose, 99+%
CAS: 3458-28-4 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 Synonym: 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,polymannose,aldehydo-d-mannose,poly mannose,mannose homopolymer,unii-pha4727wtp,aldehydo-d-manno-hexose,pha4727wtp,mannose, d,d-mannose polymers
Thermo Scientific Chemicals D-(+)-Mannose, 99%
CAS: 3458-28-4 MDL Number: MFCD00064122 Synonym: 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,polymannose,aldehydo-d-mannose,poly mannose,mannose homopolymer,unii-pha4727wtp,aldehydo-d-manno-hexose,pha4727wtp,mannose, d,d-mannose polymers
D-(+)-mannose, 100%, MP Biomedicals™
CAS: 3458-28-4 Molecular Formula: C6H12O6 Synonym: 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,polymannose,aldehydo-d-mannose,poly mannose,mannose homopolymer,unii-pha4727wtp,aldehydo-d-manno-hexose,pha4727wtp,mannose, d,d-mannose polymers
Mannose Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
D-(+)-Mannose, MP Biomedicals
CAS: 3458-28-4 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00064122 Synonym: 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,polymannose,aldehydo-d-mannose,poly mannose,mannose homopolymer,unii-pha4727wtp,aldehydo-d-manno-hexose,pha4727wtp,mannose, d,d-mannose polymers
MilliporeSigma™ Calbiochem™ D-Mannose
For Life Science reasearch application
D-(+)-Mannose 98.0+%, TCI America™
CAS: 3458-28-4 Molecular Formula: C6H12O6 MDL Number: MFCD00064122 Synonym: 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal,polymannose,aldehydo-d-mannose,poly mannose,mannose homopolymer,unii-pha4727wtp,aldehydo-d-manno-hexose,pha4727wtp,mannose, d,d-mannose polymers