Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-mannose, 100%, MP Biomedicals™

Catalog No. ICN10225080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10225080 100 g
ICN10225025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10225080 Supplier MP Biomedicals, Inc Supplier No. 0210225080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3458-28-4 | Formula: C6H12O6 | Weight: 180.16 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Color White
Density 0.35g/mL
Quantity 100 g
Molecular Weight (g/mol) 180.16
Formula Weight 180.2
Percent Purity 1
Grade Not Available
Physical Form Powder
Chemical Name or Material D-(+)-Mannose
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.