Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Mannose, MP Biomedicals

Catalog No. MP210225090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210225090 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210225090 Supplier MP Biomedicals, Inc Supplier No. 0210225090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in the formation of glycan structure and glycosylation, and can be utilized by the brain as an alternative energy source.

  • D-(+)-Mannose, a C-2 epimer of D-glucose, is used in the formation of glycan structure and glycosylation
  • D-Mannose has been used in a study to assess the synthesis of a family of amphiphilic glycopolymers
  • It has also been used in a study to investigate the early detection of bronchiolitis obliterans after lung transplantation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00064122
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Melting Point 129°C to 140°C
Color White
Boiling Point 527.1°C (760 mmHg)
Linear Formula C6H12O6
Quantity 500 g
Solubility Information Soluble in water (50 gm/mL-clear colorless solution).
Formula Weight 180.2
Percent Purity ≥98%
Grade Not Available
Physical Form Powder
Chemical Name or Material D-(+)-MANNOSE
Show More Show Less
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.