accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHCH3OOPOOPOOPOOPHClClClHRuClHHClRu(?)(?)

CAS RN 346457-41-8

IUPAC Name: [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis({4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane) hydrochloride ruthenium chloride
Synonyms: [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis(segphos) hydrochloride ruthenium chloride
Molecular Weight (g/mol): 1649.68
Molecular Formula: C78H67Cl5NO8P4Ru2
InChi Key: GTKMQYLNAZIAEF-UHFFFAOYSA-M
SMILES: Cl.[Cl-].[Ru].CNC.[ClH+][Ru-]([ClH+])[ClH+].C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1.C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)