accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NINNN(R)O(R)OH(S)HO(R)HO

CAS RN 35109-88-7

IUPAC Name: (2R,3R,4S,5R)-2-(6-amino-2-iodo-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: (2R,3R,4S,5R)-2-(6-amino-2-iodopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 393.14
Molecular Formula: C10H12IN5O4
InChi Key: MGEBVSZZNFOIRB-UUOKFMHZSA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(I)=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)