CAS RN 351344-10-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ Methyllycaconitine citrate
CAS: 351344-10-0 Molecular Formula: C43H58N2O17