accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35202-54-1

IUPAC Name: 3,4-dimethoxybicyclo[4.2.0]octa-1(6),2,4-triene-7-carbonitrile
Synonyms: 3,4-dimethoxybicyclo[4.2.0]octa-1(6),2,4-triene-7-carbonitrile
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: HJTHVTHXHHFXMJ-UHFFFAOYNA-N
SMILES: COC1=CC2=C(C=C1OC)C(C2)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results