CAS RN 353-07-1
CAS RN 353-07-1
IUPAC Name:
1-(2-cyanoethyl)hydrazine-1,2-diium
Synonyms:
1-(2-cyanoethyl)hydrazine-1,2-diium
Molecular Weight (g/mol):
87.13
Molecular Formula:
C3H9N3
InChi Key:
PDUFDGPPXZKHPU-UHFFFAOYSA-N
SMILES:
[NH3+][NH2+]CCC#N
1
–
1
of
1
results
1
2-Cyanoethylhydrazine, 98%, Thermo Scientific Chemicals
CAS: 353-07-1 Molecular Formula: C3H7N3 Molecular Weight (g/mol): 85.1 MDL Number: MFCD00043620 InChI Key: VIHRIIARIFUQLC-UHFFFAOYSA-N Synonym: 2-cyanoethylhydrazine,beta-cyanoethylhydrazine,2-cyanoethyl hydrazine,cyanoethylhydrazine,unii-l6h109c98b,3-hydrazinopropionitrile,propanenitrile, 3-hydrazino,3-hydrazinopropanenitrile PubChem CID: 136177 IUPAC Name: 3-hydrazinylpropanenitrile SMILES: C(CNN)C#N