accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO(S)NHOO

CAS RN 35661-40-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-phenylpropanoic acid|FMOC-PHE-OH
Molecular Weight (g/mol): 387.44
Molecular Formula: C24H21NO4
InChi Key: SJVFAHZPLIXNDH-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
Grade 
  • (3)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)