accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 357400-13-6

IUPAC Name: 2-(2-{[3-(1H-1,3-benzodiazol-2-yl)propyl](methyl)amino}ethyl)-6-fluoro-1-(propan-2-yl)-1,2,3,4-tetrahydronaphthalen-2-yl cyclopropanecarboxylate dihydrochloride
Synonyms: 2-(2-{[3-(1H-1,3-benzodiazol-2-yl)propyl](methyl)amino}ethyl)-6-fluoro-1-isopropyl-3,4-dihydro-1H-naphthalen-2-yl cyclopropanecarboxylate dihydrochloride
Molecular Weight (g/mol): 564.57
Molecular Formula: C30H40Cl2FN3O2
InChi Key: BCCQNBXHUMKLFW-UHFFFAOYNA-N
SMILES: Cl.Cl.CC(C)C1C2=C(CCC1(CCN(C)CCCC1=NC3=CC=CC=C3N1)OC(=O)C1CC1)C=C(F)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results