Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NNC 55-0396 dihydrochloride
GREENER_CHOICESDP

Catalog No. 226810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-681-0 Supplier Tocris Bioscience™ Supplier No. 2268/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly selective CaV3.x blocker

Highly selective T-type calcium channel blocker. Displays IC50 values of 6.8 and > 100 μM for inhibition of Cav3.1 T-type channels and HVA currents respectively in INS-1 cells.

Chemical Identifiers

CAS 357400-13-6
Molecular Formula C30H40Cl2FN3O2

Specifications

Quantity 10 mg
Formula Weight 564.57
Purity >98%
Molecular Weight (g/mol) 564.567
Chemical Name or Material NNC 55-0396 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.