accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3621-82-7

IUPAC Name: 2,6-dichloro-1,3-benzoxazole
Synonyms: 2,6-dichloro-1,3-benzoxazole
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: LVVQTPZQNHQLOM-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=C(Cl)C=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results