Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ribavirin
GREENER_CHOICESDP

Catalog No. 450150 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-015-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-015-0 Supplier R&D Systems™ Supplier No. 4501/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Serine protease inhibitor; inhibits tryptase

Antiviral guanosine ribonucleoside analog; misincorporated into mRNA by viral-dependent RNA polymerases. Binds to and redistributes mammalian eIF4E from the nucleus to the cytoplasm (Ki ∽ 0.3 μM for the active metabolite, ribavirin triphosphate). Also inosine monophosphate dehydrogenase (IMPDH) inhibitor. Represses colony formation of primary AML-M5 progenitor cells (IC50 ∽ 1 μM); reduces disease severity in acute myeloid leukemia (AML). Inhibits cytopathic effect of SARS-CoV in Vero cells in vitro. Orally available.

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 244.2
Purity >99%
Molecular Weight (g/mol) 244.207
Chemical Name or Material Protein-synthesizing GTPase Compounds
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.