accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOOF

CAS RN 368-88-7

IUPAC Name: 4-fluorobenzene-1-sulfonic acid
Synonyms: 4-fluorobenzenesulfonic acid
Molecular Weight (g/mol): 176.16
Molecular Formula: C6H5FO3S
InChi Key: WVSYONICNIDYBE-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results