accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOHONNO

CAS RN 3690-10-6

IUPAC Name: 1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: zebularine
Molecular Weight (g/mol): 228.2
Molecular Formula: C9H12N2O5
InChi Key: RPQZTTQVRYEKCR-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=CC=NC1=O
Form 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results