Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zebularine 98.0+%, TCI America™
SDP

Catalog No. Z0022200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0022200MG 200 mg
Z00221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z0022200MG Supplier TCI America Supplier No. Z0022200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3690-10-6
Molecular Formula C9H12N2O5
Molecular Weight (g/mol) 228.20
MDL Number MFCD04973699
InChI Key RPQZTTQVRYEKCR-UHFFFAOYNA-N
Synonym zebularine, pyrimidin-2-one ribonucleoside, pyrimidin-2-one beta-ribofuranoside, pyrimidin-2-one beta-d-ribofuranoside, 4-deoxyuridine, 1-beta-d-ribofuranosyl-2 1h-pyrimidinone, 1-beta-d-ribofuranosylpyrimidin-2 1h-one, unii-7a9y5sx0gy, 7a9y5sx0gy, 2 1h-pyrimidinone, 1-beta-d-ribofuranosyl
PubChem CID 100016
ChEBI CHEBI:46938
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 200 mg
Formula Weight 228.20
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Zebularine
RTECSNumber UW7539720
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.