accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNHNONOO

CAS RN 36945-98-9

IUPAC Name: 3-(2-hydroxyphenyl)-6-(3-nitrophenyl)-1,2,3,4-tetrahydropyrimidin-2-one
Synonyms: icilin
Molecular Weight (g/mol): 311.3
Molecular Formula: C16H13N3O4
InChi Key: RCEFMOGVOYEGJN-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1N1CC=C(NC1=O)C1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results