Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Icilin
GREENER_CHOICESDP

Catalog No. 153150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-315-0 50 mg
15-311-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-315-0 Supplier R&D Systems™ Supplier No. 1531/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Activates a novel cold receptor. Cooling agent

Cooling agent that activates the novel cold receptors TRPM8 (CMR1) and TRPA1 (ANKTM1/TRPN1), members of the TRP ion channel family. Induces currents in CMR1-expressing HEK 293 cells (EC50 = 0.36 μM) more potently than menthol or low temperatures. Produces "wet shakes" in vivo.

Chemical Identifiers

CAS 36945-98-9
Molecular Formula C16H13N3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 311.3
Purity >98%
Molecular Weight (g/mol) 311.297
Chemical Name or Material Icilin
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.