accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSOOFFF

CAS RN 36967-85-8

IUPAC Name: trifluoromethanesulfonyl benzoate
Synonyms: trifluoromethanesulfonyl benzoate
Molecular Weight (g/mol): 254.18
Molecular Formula: C8H5F3O4S
InChi Key: GEZVJNXOBFDIPM-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC(=O)C1=CC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results