CAS RN 375815-87-5
IUPAC Name:
(2S,3S)-2,3-dihydroxybutanedioic acid; 5,8,14-triazatetracyclo[10.3.1.0²,¹¹.0⁴,⁹]hexadeca-2(11),3,5,7,9-pentaene
Synonyms:
D-tartaric acid; varenicline
Molecular Weight (g/mol):
361.35
Molecular Formula:
C17H19N3O6
InChi Key:
TWYFGYXQSYOKLK-WUUYCOTASA-N
SMILES:
O[C@@H]([C@H](O)C(O)=O)C(O)=O.C1C2CNCC1C1=C2C=C2N=CC=NC2=C1
1
–
1
of
1
results
1
R&D Systems™ Varenicline tartrate
CAS: 375815-87-5 Molecular Formula: C17H19N3O6