accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOOOOOOOSOOCH3

CAS RN 37860-51-8

IUPAC Name: 2-[2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethoxy]ethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-[2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethoxy]ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 502.59
Molecular Formula: C22H30O9S2
InChi Key: SLAONPBUWDUSSO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results