Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylene Glycol Bis(p-toluenesulfonate) 98.0+%, TCI America™
SDP

Catalog No. T149325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T149325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. T149325G Supplier TCI America Supplier No. T149325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 37860-51-8
Molecular Formula C22H30O9S2
Molecular Weight (g/mol) 502.593
MDL Number MFCD00075239
InChI Key SLAONPBUWDUSSO-UHFFFAOYSA-N
Synonym Bis[2-[2-(p-toluenesulfonyloxy)ethoxy]ethyl] Ether, Bis[2-(2-tosyloxyethoxy)ethyl] Ether, Tetraethylene Glycol Di-p-tosylate
PubChem CID 3706987
IUPAC Name 2-[2-[2-[2-(4-methylphenyl)sulfonyloxyethoxy]ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 502.59
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tetraethylene Glycol Bis(p-toluenesulfonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.