accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38046-82-1

H3COMgBr

CAS RN 38046-82-1

IUPAC Name: bromo(6-methoxynaphthalen-2-yl)magnesium
Synonyms: bromo(6-methoxynaphthalen-2-yl)magnesium
Molecular Weight (g/mol): 261.4
Molecular Formula: C11H9BrMgO
InChi Key: SOGQGMFUEUMYJD-UHFFFAOYSA-M
SMILES: COC1=CC2=CC=C([Mg]Br)C=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results