accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHOHNNSNOONN

CAS RN 38048-32-7

IUPAC Name: 2-(hydroxymethyl)-5-(6-{[(4-nitrophenyl)methyl]sulfanyl}-9H-purin-9-yl)oxolane-3,4-diol
Synonyms: nitrobenzylthioinosine
Molecular Weight (g/mol): 419.41
Molecular Formula: C17H17N5O6S
InChi Key: DYCJFJRCWPVDHY-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=NC2=C(SCC3=CC=C(C=C3)[N+]([O-])=O)N=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)