accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClSSClClClCl

CAS RN 3808-87-5

IUPAC Name: 1,2,4-trichloro-5-[(2,4,5-trichlorophenyl)disulfanyl]benzene
Synonyms: 1,2,4-trichloro-5-[(2,4,5-trichlorophenyl)disulfanyl]benzene
Molecular Weight (g/mol): 424.98
Molecular Formula: C12H4Cl6S2
InChi Key: ZUVJVEOHQKNMPN-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(SSC2=CC(Cl)=C(Cl)C=C2Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)