accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38258-92-3

IUPAC Name: 1-methoxybicyclo[2.2.2]oct-5-ene-2-carbonitrile
Synonyms: 1-methoxybicyclo[2.2.2]oct-5-ene-2-carbonitrile
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: XQFSLPUVWRXWSQ-UHFFFAOYNA-N
SMILES: COC12CCC(CC1C#N)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results