CAS RN 38469-83-9
CAS RN 38469-83-9
IUPAC Name:
4-methoxy-2-nitrobenzonitrile
Synonyms:
4-methoxy-2-nitrobenzonitrile
Molecular Weight (g/mol):
178.15
Molecular Formula:
C8H6N2O3
InChi Key:
UUYPUFCWSUBTFP-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C#N)C(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 4-Methoxy-2-nitrobenzonitrile, 97%, Thermo Scientific™
CAS: 38469-83-9 Molecular Formula: C8H6N2O3 Molecular Weight (g/mol): 178.15 MDL Number: MFCD00667741 InChI Key: UUYPUFCWSUBTFP-UHFFFAOYSA-N Synonym: 2-nitro-4-methoxybenzonitrile,4-cyano-3-nitroanisole,4-methoxy-2-nitro-benzonitrile,benzonitrile, 4-methoxy-2-nitro,pubchem16089,acmc-209xbl,3-nitro-4-cyanoanisole,ksc580a1d,benzonitrile,4-methoxy-2-nitro,4-methoxy-2-nitrobenzonitrile PubChem CID: 288665 IUPAC Name: 4-methoxy-2-nitrobenzonitrile SMILES: COC1=CC=C(C#N)C(=C1)[N+]([O-])=O