accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClNNOO

CAS RN 38533-61-8

IUPAC Name: 2-chloro-6-methoxy-3-nitropyridine
Synonyms: 2-chloro-6-methoxy-3-nitropyridine
Molecular Weight (g/mol): 188.57
Molecular Formula: C6H5ClN2O3
InChi Key: DVRGUTNVDGIKTP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(Cl)=N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)