accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3854-04-4

IUPAC Name: trimethyl[4-(2-oxopyrrolidin-1-yl)but-3-yn-1-yl]azanium iodide
Synonyms: trimethyl[4-(2-oxopyrrolidin-1-yl)but-3-yn-1-yl]azanium iodide
Molecular Weight (g/mol): 322.19
Molecular Formula: C11H19IN2O
InChi Key: CHRKORSYUQVKAQ-UHFFFAOYSA-M
SMILES: [I-].C[N+](C)(C)CCC#CN1CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results