CAS RN 385786-48-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ HIF Prolyl Hydroxylase Inhibitor
A cell-permeable, hydroxyquinoline HIF Prolyl Hydroxylase (PHD) inhibitor that displays an IC50 = 2μM in a cell-based assay using a reporter produced by fusing HIF-1α oxygen degradable domain (ODD) to lucifer
2
Tocris Bioscience™ Adaptaquin
HIF-prolyl hydroxylase-2 (PHD2) inhibitor; antioxidant