accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39746-25-3

IUPAC Name: (5E)-7-[(1S,3S)-3-hydroxy-2-[(1E,3R)-3-hydroxy-4,4-dimethyloct-1-en-1-yl]-5-oxocyclopentyl]hept-5-enoic acid
Synonyms: (5E)-7-[(1S,3S)-3-hydroxy-2-[(1E,3R)-3-hydroxy-4,4-dimethyloct-1-en-1-yl]-5-oxocyclopentyl]hept-5-enoic acid
Molecular Weight (g/mol): 380.53
Molecular Formula: C22H36O5
InChi Key: QAOBBBBDJSWHMU-BKSAQPBESA-N
SMILES: CCCCC(C)(C)[C@H](O)\C=C\C1[C@@H](O)CC(=O)[C@H]1C\C=C\CCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results