accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40397-96-4

IUPAC Name: 1-chloro-4-isocyanato-2-nitrobenzene
Synonyms: 1-chloro-4-isocyanato-2-nitrobenzene
Molecular Weight (g/mol): 198.56
Molecular Formula: C7H3ClN2O3
InChi Key: ZCPHLSPLEGBTCZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)N=C=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)