accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNOCH3CH3HO(E)

CAS RN 404-86-4

IUPAC Name: (6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
Synonyms: capsaicin zostrix isodecenoic acid vanillylamide
Molecular Weight (g/mol): 305.42
Molecular Formula: C18H27NO3
InChi Key: YKPUWZUDDOIDPM-SOFGYWHQSA-N
SMILES: COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O
Grade 
  • (2)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (5)
Physical Form 
  • (1)
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)