CAS RN 405060-95-9
IUPAC Name:
3-phenyl-N-(2,2,2-trichloro-1-{[(quinolin-8-yl)carbamothioyl]amino}ethyl)prop-2-enamide
Synonyms:
salubrinal
Molecular Weight (g/mol):
479.8
Molecular Formula:
C21H17Cl3N4OS
InChi Key:
LCOIAYJMPKXARU-UHFFFAOYNA-N
SMILES:
ClC(Cl)(Cl)C(NC(=S)NC1=C2N=CC=CC2=CC=C1)NC(=O)C=CC1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Salubrinal
Thermo Scientific Chemicals Salubrinal
CAS: 405060-95-9 Molecular Formula: C21H17Cl3N4OS Synonym: eIF-2alpha Inhibitor; SAL
2
R&D Systems™ Salubrinal
CAS: 405060-95-9 Molecular Formula: C21H17Cl3N4OS