CAS RN 405098-33-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ C5a Receptor Antagonist, W-54011
The C5a Receptor Antagonist, W-54011, also referenced under CAS 405098-33-1, controls the biological activity of C5a Receptor.
2
Tocris Bioscience™ W 54011
High affinity C5a receptor antagonist