accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNNH2NNH(E/Z)

CAS RN 406208-42-2

IUPAC Name: 2-amino-6-[2-(cyclopropylmethoxy)-6-oxocyclohexa-2,4-dien-1-ylidene]-4-(piperidin-4-yl)-1,6-dihydropyridine-3-carbonitrile
Synonyms: 2-amino-6-[2-(cyclopropylmethoxy)-6-oxocyclohexa-2,4-dien-1-ylidene]-4-(piperidin-4-yl)-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 364.45
Molecular Formula: C21H24N4O2
InChi Key: DWQVGCQPWXKRKO-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C(=CC(N1)=C1C(=O)C=CC=C1OCC1CC1)C1CCNCC1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results