accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNH

CAS RN 4079-26-9

IUPAC Name: 6H,11H-thiochromeno[4,3-b]indole
Synonyms: 6H,11H-thiochromeno[4,3-b]indole
Molecular Weight (g/mol): 237.32
Molecular Formula: C15H11NS
InChi Key: ZGOOPZVQMLHPFM-UHFFFAOYSA-N
SMILES: C1SC2=CC=CC=C2C2=C1C1=CC=CC=C1N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)