Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PD 146176
GREENER_CHOICESDP

Catalog No. 285050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-505-0 50 mg
28-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-505-0 Supplier R&D Systems™ Supplier No. 2850/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective 15-lipoxygenase inhibitor

Specific, non-competitve 15-lipoxygenase (15-LOX) inhibitor (Ki = 197nM) that has no demonstrable effect on 5-LOX, 12-LOX, COX-1 or COX-2 (IC50 = 0.54 μM for 15-LOX in rabbit reticulocytes). Lacks non-specific antioxidant properties and prevents atherogenesis via regulation of monocyte-macrophage enrichment in vivo.

Chemical Identifiers

CAS 4079-26-9
Molecular Formula C15H11NS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 237.32
Purity >98%
Molecular Weight (g/mol) 237.32
Chemical Name or Material PD 146176
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.