CAS RN 41100-52-1
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%
CAS: 41100-52-1 Molecular Formula: C12H22ClN Molecular Weight (g/mol): 215.77 MDL Number: MFCD00214336 InChI Key: LDDHMLJTFXJGPI-UHFFFAOYNA-N Synonym: memantine hydrochloride,memantine hcl,3,5-dimethyladamantan-1-amine hydrochloride,3,5-dimethyl-1-adamantanamine hydrochloride,namenda,akatinol,axura,namenda xr,memantine.hcl PubChem CID: 181458 ChEBI: CHEBI:64323 IUPAC Name: 3,5-dimethyladamantan-1-amine;hydrochloride SMILES: [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
Memantine hydrochloride
CAS: 41100-52-1 Molecular Formula: C12H22ClN Molecular Weight (g/mol): 215.77 MDL Number: MFCD00214336 InChI Key: LDDHMLJTFXJGPI-UHFFFAOYNA-N Synonym: memantine hydrochloride,memantine hcl,3,5-dimethyladamantan-1-amine hydrochloride,3,5-dimethyl-1-adamantanamine hydrochloride,namenda,akatinol,axura,namenda xr,memantine.hcl PubChem CID: 181458 ChEBI: CHEBI:64323 IUPAC Name: hydrogen 3,5-dimethyladamantan-1-amine chloride SMILES: [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
3,5-Dimethyl-1-adamantanamine Hydrochloride 98.0+%, TCI America™
CAS: 41100-52-1 Molecular Formula: C12H22ClN Molecular Weight (g/mol): 215.77 MDL Number: MFCD00214336 InChI Key: LDDHMLJTFXJGPI-UHFFFAOYNA-N Synonym: memantine hydrochloride,memantine hcl,3,5-dimethyladamantan-1-amine hydrochloride,3,5-dimethyl-1-adamantanamine hydrochloride,namenda,akatinol,axura,namenda xr,memantine.hcl PubChem CID: 181458 ChEBI: CHEBI:64323 IUPAC Name: hydrogen 3,5-dimethyladamantan-1-amine chloride SMILES: [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
R&D Systems™ Memantine hydrochloride
CAS: 41100-52-1 Molecular Formula: C12H22ClN