accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 4113-12-6

IUPAC Name: 1-(hexadecyloxy)hexadecane
Synonyms: dicetyl ether
Molecular Weight (g/mol): 466.88
Molecular Formula: C32H66O
InChi Key: FDCJDKXCCYFOCV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOCCCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results