accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNI

CAS RN 41270-96-6

IUPAC Name: 4-iodo-6-phenylpyrimidine
Synonyms: 4-iodo-6-phenylpyrimidine
Molecular Weight (g/mol): 282.08
Molecular Formula: C10H7IN2
InChi Key: ZTCJXHNJVLUUMR-UHFFFAOYSA-N
SMILES: IC1=NC=NC(=C1)C1=CC=CC=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results