Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MIF Antagonist III, 4-IPP
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
Description
A cell-permeable iodo-pyrimidine compound that covalently modifies MIF N-terminal proline and exhibits higher MIF inhibitory potency than ISO-1 (Cat. No. 475837) in cell-free dopachrome tautomerase activity assays (IC50 ∽5 vs. 50μM, respectively, using recombinant human MIF) as well as in cell-based migration assays (50% vs 0% inhibition with respective inhibitor at 10μM) and anchorage-independent growth (57% vs 0% inhibition with respective inhibitor at 10μM) using human lung adenocarcinoma A549. Also shown to reduce A549 cellular active Rac1 population (50μM, 16 h) in vitro and effectively decrease liver tautomerase activity by 50% in mice (1mg/animal/day or ∽50mg/kg/day via i.p.) in vivo .

Specifications
Specifications
Color | Amber |
Description | 90% by HPLC |
CAS | 41270-96-6 |
Molecular Formula | C10H7IN2 |
Formula Weight | 282.1g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction