CAS RN 423768-52-9
CAS RN 423768-52-9
IUPAC Name:
1-(1,5-dimethyl-1H-pyrazol-3-yl)methanamine
Synonyms:
1-(1,5-dimethylpyrazol-3-yl)methanamine
Molecular Weight (g/mol):
125.18
Molecular Formula:
C6H11N3
InChi Key:
JGYXJOBBROGMLL-UHFFFAOYSA-N
SMILES:
CN1N=C(CN)C=C1C
1
–
1
of
1
results
1
Thermo Scientific Chemicals (1,5-Dimethyl-1H-pyrazol-3-yl)methylamine, 97%, Thermo Scientific™
CAS: 423768-52-9 Molecular Formula: C6H11N3 Molecular Weight (g/mol): 125.18 MDL Number: MFCD03422531 InChI Key: JGYXJOBBROGMLL-UHFFFAOYSA-N Synonym: 1,5-dimethyl-1h-pyrazol-3-yl methanamine,1,5-dimethyl-1h-pyrazol-3-yl methylamine,3-aminomethyl-1,5-dimethylpyrazole,1-1,5-dimethyl-1h-pyrazol-3-yl methanamine,1,5-dimethylpyrazol-3-yl methanamine,3-aminomethyl-1,5-dimethyl-1h-pyrazole,1,5-dimethylpyrazol-3-yl methylamine,1h-pyrazole-3-methanamine,1,5-dimethyl,1-1,5-dimethylpyrazol-3-yl methanamine,1,5-dimethyl-1h-pyrazol-3-yl methanamine 2hcl PubChem CID: 2776367 SMILES: CN1N=C(CN)C=C1C